missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-tert-Leucine Methyl Ester Hydrochloride, TCI America™

$39.72 - $378.89

Chemical Identifiers

CAS 63038-27-7
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
InChI Key HRTQWUHFSXVRPY-NUBCRITNSA-N
Synonym l-tert-leucine methyl ester hydrochloride, h-tle-ome.hcl, s-methyl 2-amino-3,3-dimethylbutanoate hydrochloride, methyl 2s-2-amino-3,3-dimethylbutanoate hydrochloride, s-2-amino-3,3-dimethylbutyric acid methyl ester hydrochloride, methyl 3-methyl-l-valinate hydrochloride, pubchem11076, methyl l-tert-leucinate hydrochloride, l-tert-leucine methyl ester hcl, l-tert-leucine methyl-ester hydrochloride
PubChem CID 16217630
IUPAC Name methyl (2S)-2-amino-3,3-dimethylbutanoate;hydrochloride
SMILES CC(C)(C)C(C(=O)OC)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L01881G View Documents TCI America
L01881G
1g Each for $39.72
 
L018825G View Documents TCI America
L018825G
25g Each for $378.89
 
L01885G View Documents TCI America
L01885G
5g Each for $126.81
 
Specifications

Chemical Identifiers

63038-27-7
181.66
l-tert-leucine methyl ester hydrochloride, h-tle-ome.hcl, s-methyl 2-amino-3,3-dimethylbutanoate hydrochloride, methyl 2s-2-amino-3,3-dimethylbutanoate hydrochloride, s-2-amino-3,3-dimethylbutyric acid methyl ester hydrochloride, methyl 3-methyl-l-valinate hydrochloride, pubchem11076, methyl l-tert-leucinate hydrochloride, l-tert-leucine methyl ester hcl, l-tert-leucine methyl-ester hydrochloride
methyl (2S)-2-amino-3,3-dimethylbutanoate;hydrochloride
C7H16ClNO2
HRTQWUHFSXVRPY-NUBCRITNSA-N
16217630
CC(C)(C)C(C(=O)OC)N.Cl

Specifications

White
63038-27-7
l-tert-leucine methyl ester hydrochloride, h-tle-ome.hcl, s-methyl 2-amino-3,3-dimethylbutanoate hydrochloride, methyl 2s-2-amino-3,3-dimethylbutanoate hydrochloride, s-2-amino-3,3-dimethylbutyric acid methyl ester hydrochloride, methyl 3-methyl-l-valinate hydrochloride, pubchem11076, methyl l-tert-leucinate hydrochloride, l-tert-leucine methyl ester hcl, l-tert-leucine methyl-ester hydrochloride
CC(C)(C)C(C(=O)OC)N.Cl
181.66
181.66
≥98.0% (N)
1g
C7H16ClNO2
HRTQWUHFSXVRPY-NUBCRITNSA-N
methyl (2S)-2-amino-3,3-dimethylbutanoate;hydrochloride
16217630
Crystal-Powder at 20°C
L-tert-Leucine Methyl Ester Hydrochloride
Safety and Handling

Safety and Handling

TSCA : No