Please login to your online account to display your discounted pricing

L-(+)-Tartaric Acid (Fine Granular/NF/FCC), Fisher Chemical

$302.43 - $2,550.87

Chemical Identifiers

Chemical Name or Material L-(+)-Tartaric Acid
CAS 87-69-4
Purity Grade FCC/NF
Physical Form Solid
Assay 99.7 to 100.5% (NF)
Molecular Formula C4H6O6
Linear Formula HOOC(CHOH)2COOH
Formula Weight 150.09
MDL Number MFCD00064207
Synonym 2, 3-Dihydroxybutanedioic Acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A313-500

  SDS
fisher scientific company
FLA313500
Poly Bottle 500g Each for $302.43
Case of 6 Each for $1,814.58

A313-12

  SDS
thermo fisher scientific
FLA31312
Poly Pail 12kg Each for $2,550.87
Description & Specifications

Specifications

Melting Point 168°C
Purity Grade FCC/NF
Formula Weight 150.09
Vapor Pressure <0.1 mbar at 20°C
Quantity 500g
Alpha Vector Tartaric Acid
Physical Form Solid
Assay 99.7 to 100.5% (NF)
pH 1.6
Color White
Specific Rotation 12.0 to 13.0° (FCC)
Sulfate Pass Test (FCC)
Synonym 2, 3-Dihydroxybutanedioic Acid
Identification Pass Test (FCC)
Ignition Residue 0.05% max. (FCC)
Loss on Drying 0.5% max. (FCC)
Oxalate Pass Test (FCC)
Residue after Ignition 0.1% max. (NF)
MDL Number MFCD00064207
Heavy Metals (as Pb) 0.001% max. (NF)
Lead (Pb) 2mg/kg max. (FCC)
Packaging Poly Bottle
Residual Solvents Meets Requirements (NF)
Linear Formula HOOC(CHOH)2COOH
ChemAlert Storage Symbol Gray
Chemical Name or Material L-(+)-Tartaric Acid
Molecular Formula C4H6O6
CAS 87-69-4

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling