Please login to your online account to display your discounted pricing

L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC424441000

 View more versions of this product

Catalog No. AC424441000

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Needles or Powder
Formula Weight 230.08
Quantity 100g
Color White
Specific Rotation Condition +26.50° (20°C c=1,water)
pH 7.0 to 9.0 (5% soln. at 25°C)
Assay 99.0 to 101.0%
Chemical Name or Material L+-Tartaric acid, disodium salt, dihydrate
Sulfate 50ppm max.
Ammonium 30ppm max.
Calcium (Ca) 100ppm max.
Assay Percent Range 99+%
Chlorine (Cl) 5ppm max.
Molecular Formula C4H4Na2O6·2H2O
Synonym Disodium tartrate
Heavy Metals (as Pb) 5ppm max.
Infrared Spectrum Authentic
Insoluble Matter 0.005% max.
Iron (Fe) 10ppm max.
Loss on Drying 15.61% to 15.71% (150°C)
Packaging Plastic bottle
Phosphate 5ppm max.
Linear Formula (CHOHCOONa)2·2H2O
MDL Number MFCD00150035
CAS 6106-24-7

Specifications

Chemical Name or Material L+-Tartaric acid, disodium salt, dihydrate
CAS 6106-24-7
Purity Grade ACS Reagent Grade
Assay 99.0 to 101.0%
Assay Percent Range 99+%
Molecular Formula C4H4Na2O6·2H2O
Linear Formula (CHOHCOONa)2·2H2O
Formula Weight 230.08
MDL Number MFCD00150035
Synonym Disodium tartrate