Please login to your online account to display your discounted pricing

L-(+)-Tartaric acid dipotassium salt, 97%, Acros Organics


Quantity 100g

Manufacturer: thermo fisher scientific  AC427991000

 View more versions of this product

Catalog No. AC427991000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 226.27
Quantity 100g
Merck Index 15, 7796
CAS 921-53-9
Chemical Name or Material L-(+)-Tartaric acid dipotassium salt
Molecular Formula C4H4K2O6
Packaging Plastic bottle
Assay Percent Range 97%

Specifications

Chemical Name or Material L-(+)-Tartaric acid dipotassium salt
Assay Percent Range 97%
Molecular Formula C4H4K2O6
Formula Weight 226.27