missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L(-)-Sorbose, 98%, ACROS Organics™
$450.93 - $2,646.09
Chemical Identifiers
CAS | 87-79-6 |
---|---|
Molecular Formula | C6H12O6 |
Molecular Weight (g/mol) | 180.16 |
MDL Number | MFCD00151097 |
InChI Key | BJHIKXHVCXFQLS-OTWZMJIISA-N |
Synonym | 3s,4r,5s-1,3,4,5,6-pentahydroxyhexan-2-one, esorben, hsdb 780, keto-l-sorbose, l---sorbose, l--sorbose, l-sorbinose, l-xylo-2-hexulose, sorbin, sorbinose |
PubChem CID | 6904 |
ChEBI | CHEBI:13172 |
IUPAC Name | (3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one |
SMILES | C(C(C(C(C(=O)CO)O)O)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC225961000
|
Thermo Fisher Scientific
AC225961000 |
100g | Plastic bottle |
Each for $450.93
|
|
AC225962500
|
Thermo Fisher Scientific
AC225962500 |
250g | Plastic bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC225960010
|
Thermo Fisher Scientific
AC225960010 |
1kg | Plastic bottle |
Each for $2,646.09
|
|
Chemical Identifiers
87-79-6 | |
180.16 | |
BJHIKXHVCXFQLS-OTWZMJIISA-N | |
6904 | |
(3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one |
C6H12O6 | |
MFCD00151097 | |
3s,4r,5s-1,3,4,5,6-pentahydroxyhexan-2-one, esorben, hsdb 780, keto-l-sorbose, l---sorbose, l--sorbose, l-sorbinose, l-xylo-2-hexulose, sorbin, sorbinose | |
CHEBI:13172 | |
C(C(C(C(C(=O)CO)O)O)O)O |
Specifications
Authentic | |
Plastic bottle | |
-43 | |
158°C to 160°C | |
87-79-6 | |
C6H12O6 | |
15,8852 | |
Solubility in water: soluble. Other solubilities: almost insoluble in alcohol,slightly soluble in ethanol,ethylether,methanol | |
C(C(C(C(C(=O)CO)O)O)O)O | |
180.16 | |
CHEBI:13172 | |
Crystalline Powder or Crystals and Chunks | |
L(-)-Sorbose, 99% |
1% max. | |
- 43.00 (20.00°C c=5,H2O,23 C) | |
White | |
100g | |
97.5% min. (ELSD) (HPLC) | |
MFCD00151097 | |
3s,4r,5s-1,3,4,5,6-pentahydroxyhexan-2-one, esorben, hsdb 780, keto-l-sorbose, l---sorbose, l--sorbose, l-sorbinose, l-xylo-2-hexulose, sorbin, sorbinose | |
BJHIKXHVCXFQLS-OTWZMJIISA-N | |
(3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one | |
6904 | |
180.16 | |
98% |
Safety and Handling
EINECSNumber : 201-771-8
RTECSNumber : WG3195025
TSCA : TSCA