missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(-)-Sorbose, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC225962500

 View more versions of this product

Catalog No. AC225962500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

L(-)-Sorbose
1% max.
- 43.00 (20.00°C c=5,H2O,23 C)
White
250g
87-79-6
C6H12O6
15,8852
Solubility in water: soluble. Other solubilities: almost insoluble in alcohol,slightly soluble in ethanol,ethylether,methanol
C(C(C(C(C(=O)CO)O)O)O)O
180.16
CHEBI:13172
Crystalline Powder or Crystals and Chunks
Authentic
Plastic bottle
-43
158°C to 160°C
99%
97.5% min. (ELSD) (HPLC)
MFCD00151097
3s,4r,5s-1,3,4,5,6-pentahydroxyhexan-2-one, esorben, hsdb 780, keto-l-sorbose, l---sorbose, l--sorbose, l-sorbinose, l-xylo-2-hexulose, sorbin, sorbinose
BJHIKXHVCXFQLS-OTWZMJIISA-N
(3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one
6904
180.16
98%

Chemical Identifiers

87-79-6
180.16
BJHIKXHVCXFQLS-OTWZMJIISA-N
6904
(3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one
C6H12O6
MFCD00151097
3s,4r,5s-1,3,4,5,6-pentahydroxyhexan-2-one, esorben, hsdb 780, keto-l-sorbose, l---sorbose, l--sorbose, l-sorbinose, l-xylo-2-hexulose, sorbin, sorbinose
CHEBI:13172
C(C(C(C(C(=O)CO)O)O)O)O
Safety and Handling

Safety and Handling

EINECSNumber : 201-771-8

RTECSNumber : WG3195025

TSCA : TSCA

SDS Documents