missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Serine benzyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2879003
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
∽175°C (decomposition) | |
60022-62-0 | |
MFCD00038955 | |
Soluble in water. | |
Cl.NC(CO)C(=O)OCC1=CC=CC=C1 | |
231.68 | |
231.68 | |
1 g |
L-Serine benzyl ester hydrochloride | |
C10H14ClNO3 | |
l-serine benzyl ester hydrochloride, h-ser-obzl.hcl, ser bzl .hcl, h-ser.obzl.hcl, benzyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, phenylmethyl ester, hydrochloride, l-serinebenzylesterhydrochloride, h-ser-obzl. hcl, pubchem19043, h-ser-obn hydrochloride | |
MGZWCDQAKCHOBX-UHFFFAOYNA-N | |
benzyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride | |
12930233 | |
98% | |
−12° (c=4 in Water) |
Chemical Identifiers
60022-62-0 | |
231.68 | |
MGZWCDQAKCHOBX-UHFFFAOYNA-N | |
12930233 |
C10H14ClNO3 | |
MFCD00038955 | |
l-serine benzyl ester hydrochloride, h-ser-obzl.hcl, ser bzl .hcl, h-ser.obzl.hcl, benzyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, phenylmethyl ester, hydrochloride, l-serinebenzylesterhydrochloride, h-ser-obzl. hcl, pubchem19043, h-ser-obn hydrochloride | |
Cl.NC(CO)C(=O)OCC1=CC=CC=C1 |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only