missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Serine, ≥99%, MP Biomedicals™
$83.81 - $83.81
Chemical Identifiers
CAS | 56-45-1 |
---|---|
Molecular Formula | C3H7NO3 |
Molecular Weight (g/mol) | 105.093 |
MDL Number | MFCD00064224 |
InChI Key | MTCFGRXMJLQNBG-REOHCLBHSA-N |
Synonym | 2s-2-amino-3-hydroxypropanoic acid, beta-hydroxyalanine, h-ser-oh, l---serine, l-ser, l-serine, s-2-amino-3-hydroxypropanoic acid, s-serine, serine, serine, l |
PubChem CID | 5951 |
ChEBI | CHEBI:17115 |
IUPAC Name | (2S)-2-amino-3-hydroxypropanoic acid |
SMILES | C(C(C(=O)O)N)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10287325
|
Mp Biomedicals
0210287325 |
25g |
Each for $83.81
|
|
Chemical Identifiers
56-45-1 | |
105.093 | |
MTCFGRXMJLQNBG-REOHCLBHSA-N | |
5951 | |
(2S)-2-amino-3-hydroxypropanoic acid |
C3H7NO3 | |
MFCD00064224 | |
2s-2-amino-3-hydroxypropanoic acid, beta-hydroxyalanine, h-ser-oh, l---serine, l-ser, l-serine, s-2-amino-3-hydroxypropanoic acid, s-serine, serine, serine, l | |
CHEBI:17115 | |
C(C(C(=O)O)N)O |
Specifications
56-45-1 | |
C3H7NO3 | |
2s-2-amino-3-hydroxypropanoic acid, beta-hydroxyalanine, h-ser-oh, l---serine, l-ser, l-serine, s-2-amino-3-hydroxypropanoic acid, s-serine, serine, serine, l | |
C(C(C(=O)O)N)O | |
105.093 | |
CHEBI:17115 | |
Free-Flowing Powder | |
25g |
≥99% | |
MFCD00064224 | |
MTCFGRXMJLQNBG-REOHCLBHSA-N | |
(2S)-2-amino-3-hydroxypropanoic acid | |
5951 | |
105.09 | |
White | |
L-Serine |
Safety and Handling
EINECSNumber : 200-274-3
Recommended Storage : Store at room temperature.