missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Serine, ≥99%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0210287325
Specifications
L-Serine | |
≥99% | |
MFCD00064224 | |
MTCFGRXMJLQNBG-REOHCLBHSA-N | |
(2S)-2-amino-3-hydroxypropanoic acid | |
5951 | |
105.09 | |
White |
56-45-1 | |
C3H7NO3 | |
2s-2-amino-3-hydroxypropanoic acid, beta-hydroxyalanine, h-ser-oh, l---serine, l-ser, l-serine, s-2-amino-3-hydroxypropanoic acid, s-serine, serine, serine, l | |
C(C(C(=O)O)N)O | |
105.093 | |
CHEBI:17115 | |
Free-Flowing Powder | |
25g |
Chemical Identifiers
56-45-1 | |
105.093 | |
MTCFGRXMJLQNBG-REOHCLBHSA-N | |
5951 | |
(2S)-2-amino-3-hydroxypropanoic acid |
C3H7NO3 | |
MFCD00064224 | |
2s-2-amino-3-hydroxypropanoic acid, beta-hydroxyalanine, h-ser-oh, l---serine, l-ser, l-serine, s-2-amino-3-hydroxypropanoic acid, s-serine, serine, serine, l | |
CHEBI:17115 | |
C(C(C(=O)O)N)O |
Safety and Handling
EINECSNumber : 200-274-3
Recommended Storage : Store at room temperature.