Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Selectin, Certified Mass Spec grade


For research use

Manufacturer: alfa aesar™  J67853ODL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67853ODL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 3 vials
Recombinant No
Storage Requirements -30° to -10°C
Regulatory Status RUO
Product Type L-Selectin, Certified Mass Spec grade
Format Lyophilized powder
Formulation Lyophilized powder