missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-(+)-Rhamnose Monohydrate 98.0+%, TCI America™

Manufacturer:  Tci America R001325G

 View more versions of this product

Catalog No. R001325G



Specifications

Specifications

L-(+)-Rhamnose Monohydrate
93°C
10030-85-0
MFCD00149363
CBDCDOTZPYZPRO-DEZHIRTDSA-N
(2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate
20849066
Crystal-Powder at 20°C
White
25g
C6H14O6
l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
CC(C(C(C(C=O)O)O)O)O.O
182.172
164.16
≥98.0% (HPLC)

Chemical Identifiers

10030-85-0
182.172
CBDCDOTZPYZPRO-DEZHIRTDSA-N
20849066
CC(C(C(C(C=O)O)O)O)O.O
C6H14O6
MFCD00149363
l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
(2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents