missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Rhamnose Monohydrate, Promega™ DFS Item

Reagent used to induce protein expression from vectors with ORFs expressed from T7 RNA polymerase and transformed into Single Step (KRX) E. coli cells.

Manufacturer:  Promega Corportaion L5701

 View more versions of this product

Catalog No. PRL5701



Description

Description

For molecular biology, cloning applications
Specifications

Specifications

L-Rhamnose Monohydrate
10g

Chemical Identifiers

0.0
icon-pdf.png
Safety and Handling

Safety and Handling

Recommended Storage : 15 to 30°C

Documents
Promotions

Promotions