missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-(+)-Rhamnose, monohydrate, ∽99%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0210280910
Specifications
L-(+)-Rhamnose, monohydrate | |
White | |
10030-85-0 | |
C6H14O6 | |
2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, 6-deoxy-l-mannose hydrate, a-l-rhamnose monohydrate, l + rhamnopyranose, l +-rhamnose hydrate, l-+-rhamnose monohydrate, l-mannose, 6-deoxy-, monohydrate, l-rha hydrate, l-rhamnose hydrate, rhamnose hydrate | |
CC(C(C(C(C=O)O)O)O)O.O | |
182.172 | |
182.17 | |
∽99% |
1.47g/mL | |
10g | |
∽ 99% | |
MFCD00071591 | |
CBDCDOTZPYZPRO-DEZHIRTDSA-N | |
(2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate | |
20849066 | |
Solid |
Chemical Identifiers
10030-85-0 | |
182.172 | |
CBDCDOTZPYZPRO-DEZHIRTDSA-N | |
20849066 | |
CC(C(C(C(C=O)O)O)O)O.O |
C6H14O6 | |
MFCD00071591 | |
2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, 6-deoxy-l-mannose hydrate, a-l-rhamnose monohydrate, l + rhamnopyranose, l +-rhamnose hydrate, l-+-rhamnose monohydrate, l-mannose, 6-deoxy-, monohydrate, l-rha hydrate, l-rhamnose hydrate, rhamnose hydrate | |
(2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate |
Safety and Handling
Recommended Storage : Store at room temperature.