missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Proline tert-butyl ester, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438110010
Specifications
L-Proline tert-butyl ester | |
Glass bottle | |
50°C to 51°C (1.0 Torr) | |
1g | |
96% min. (GC) | |
h-pro-otbu, l-proline t-butyl ester, l-proline t-butyl ester free base, l-proline tert-butyl ester, l-proline, 1,1-dimethylethyl ester, l-proline-t-butyl ester, s-tert-butyl pyrrolidine-2-carboxylate, t-butyl prolinate, tert-butyl 2s-pyrrolidine-2-carboxylate, tert-butyl l-prolinate | |
CC(C)(C)OC(=O)C1CCCN1 | |
171.24 | |
171.24 | |
97% |
Authentic | |
-40° to -48° (20°C, 589nm) (c=1, ethanol) | |
Colorless to Yellow | |
2812-46-6 | |
C9H17NO2 | |
XJJBXZIKXFOMLP-ZETCQYMHSA-N | |
tert-butyl (2S)-pyrrolidine-2-carboxylate | |
6453108 | |
Liquid |
Chemical Identifiers
2812-46-6 | |
171.24 | |
h-pro-otbu, l-proline t-butyl ester, l-proline t-butyl ester free base, l-proline tert-butyl ester, l-proline, 1,1-dimethylethyl ester, l-proline-t-butyl ester, s-tert-butyl pyrrolidine-2-carboxylate, t-butyl prolinate, tert-butyl 2s-pyrrolidine-2-carboxylate, tert-butyl l-prolinate | |
tert-butyl (2S)-pyrrolidine-2-carboxylate |
C9H17NO2 | |
XJJBXZIKXFOMLP-ZETCQYMHSA-N | |
6453108 | |
CC(C)(C)OC(=O)C1CCCN1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
EINECSNumber : 220-558-