missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Proline tert-butyl ester, 97%, ACROS Organics™

Chemical Identifiers

CAS 2812-46-6
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
InChI Key XJJBXZIKXFOMLP-ZETCQYMHSA-N
Synonym h-pro-otbu, l-proline t-butyl ester, l-proline t-butyl ester free base, l-proline tert-butyl ester, l-proline, 1,1-dimethylethyl ester, l-proline-t-butyl ester, s-tert-butyl pyrrolidine-2-carboxylate, t-butyl prolinate, tert-butyl 2s-pyrrolidine-2-carboxylate, tert-butyl l-prolinate
PubChem CID 6453108
IUPAC Name tert-butyl (2S)-pyrrolidine-2-carboxylate
SMILES CC(C)(C)OC(=O)C1CCCN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC438110010
SDS View Documents
Thermo Fisher Scientific
AC438110010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC438110050
SDS View Documents
Thermo Fisher Scientific
AC438110050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

2812-46-6
171.24
h-pro-otbu, l-proline t-butyl ester, l-proline t-butyl ester free base, l-proline tert-butyl ester, l-proline, 1,1-dimethylethyl ester, l-proline-t-butyl ester, s-tert-butyl pyrrolidine-2-carboxylate, t-butyl prolinate, tert-butyl 2s-pyrrolidine-2-carboxylate, tert-butyl l-prolinate
tert-butyl (2S)-pyrrolidine-2-carboxylate
C9H17NO2
XJJBXZIKXFOMLP-ZETCQYMHSA-N
6453108
CC(C)(C)OC(=O)C1CCCN1

Specifications

Authentic
-40° to -48° (20°C, 589nm) (c=1, ethanol)
Colorless to Yellow
2812-46-6
C9H17NO2
XJJBXZIKXFOMLP-ZETCQYMHSA-N
tert-butyl (2S)-pyrrolidine-2-carboxylate
6453108
Liquid
L-Proline tert-butyl ester
Glass bottle
50°C to 51°C (1.0 Torr)
1g
96% min. (GC)
h-pro-otbu, l-proline t-butyl ester, l-proline t-butyl ester free base, l-proline tert-butyl ester, l-proline, 1,1-dimethylethyl ester, l-proline-t-butyl ester, s-tert-butyl pyrrolidine-2-carboxylate, t-butyl prolinate, tert-butyl 2s-pyrrolidine-2-carboxylate, tert-butyl l-prolinate
CC(C)(C)OC(=O)C1CCCN1
171.24
171.24
97%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

EINECSNumber : 220-558-

SDS