Please login to your online account to display your discounted pricing

L-Proline benzyl ester hydrochloride, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC320600010

 View more versions of this product

Catalog No. AC320600010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay 97.5 to 102.5% (Argentometry), ≥98.5% (HPLC)
Physical Form Crystalline Powder
Formula Weight 241.72
Quantity 1g
Color White to pale yellow
EINECS Number 240-700-5
Specific Rotation Condition −44° (20°C c=1,methanol)
Chemical Name or Material L-Proline benzyl ester hydrochloride
Name Note 98%
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C12H15NO2·ClH
Packaging Glass bottle
MDL Number MFCD00039002
CAS 16652-71-4
Specific Rotation -43° to -45° (20°C, 589nm) (c=1, methanol)
Health Hazard 1 HYGROSCOPIC
Water 0.5% max. (K.F.)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Additional Information D-enantiomer: < 0.5%
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material L-Proline benzyl ester hydrochloride
Name Note 98%
CAS 16652-71-4
Assay 97.5 to 102.5% (Argentometry), ≥98.5% (HPLC)
Assay Percent Range 98%
Molecular Formula C12H15NO2·ClH
Formula Weight 241.72
MDL Number MFCD00039002
Accessories

HYGROSCOPIC

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents