Please login to your online account to display your discounted pricing

L-Proline benzyl ester hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material L-Proline benzyl ester hydrochloride
Name Note 98%
CAS 16652-71-4
Assay 97.5 to 102.5% (Argentometry), ≥98.5% (HPLC)
Assay Percent Range 98%
Molecular Formula C12H15NO2·ClH
Formula Weight 241.72
MDL Number MFCD00039002
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC320600010

  SDS
thermo fisher scientific
AC320600010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC320600050

  SDS
fisher scientific acros
320600050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay 97.5 to 102.5% (Argentometry), ≥98.5% (HPLC)
Physical Form Crystalline Powder
Formula Weight 241.72
Quantity 1g
Color White to pale yellow
EINECS Number 240-700-5
Specific Rotation Condition −44° (20°C c=1,methanol)
Specific Rotation -43° to -45° (20°C, 589nm) (c=1, methanol)
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Water 0.5% max. (K.F.)
Infrared Spectrum Authentic
Additional Information D-enantiomer: < 0.5%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 HYGROSCOPIC
Name Note 98%
Molecular Formula C12H15NO2·ClH
CAS 16652-71-4
Assay Percent Range 98%
Chemical Name or Material L-Proline benzyl ester hydrochloride
MDL Number MFCD00039002
Safety and Handling

HYGROSCOPIC

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.