missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Phenylephrine 98.0+%, TCI America™
$164.17 - $494.08
Chemical Identifiers
CAS | 59-42-7 |
---|---|
Molecular Formula | C9H13NO2 |
Molecular Weight (g/mol) | 167.208 |
MDL Number | MFCD00044749 |
InChI Key | SONNWYBIRXJNDC-VIFPVBQESA-N |
Synonym | (R)-(-)-Phenylephrine, (R)-(-)-3-[1-Hydroxy-2-(methylamino)ethyl]phenol, (R)-(-)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol, (R)-(-)-3-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol |
PubChem CID | 6041 |
ChEBI | CHEBI:8093 |
IUPAC Name | 3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol |
SMILES | CNCC(C1=CC(=CC=C1)O)O |
Chemical Identifiers
59-42-7 | |
167.208 | |
SONNWYBIRXJNDC-VIFPVBQESA-N | |
6041 | |
3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol |
C9H13NO2 | |
MFCD00044749 | |
(R)-(-)-Phenylephrine, (R)-(-)-3-[1-Hydroxy-2-(methylamino)ethyl]phenol, (R)-(-)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol, (R)-(-)-3-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol | |
CHEBI:8093 | |
CNCC(C1=CC(=CC=C1)O)O |
Specifications
White-Yellow | |
5g | |
C9H13NO2 | |
(R)-(-)-Phenylephrine, (R)-(-)-3-[1-Hydroxy-2-(methylamino)ethyl]phenol, (R)-(-)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol, (R)-(-)-3-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol | |
CNCC(C1=CC(=CC=C1)O)O | |
167.208 | |
CHEBI:8093 | |
Crystal-Powder at 20°C | |
L-Phenylephrine |
174°C | |
59-42-7 | |
MFCD00044749 | |
SONNWYBIRXJNDC-VIFPVBQESA-N | |
3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol | |
6041 | |
167.21 | |
≥98.0% (HPLC,T) |
Safety and Handling
RTECSNumber : DO7175000
TSCA : No