Please login to your online account to display your discounted pricing

L(-)-Phenylephrine hydrochloride, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC207241000

 View more versions of this product

Catalog No. AC207241000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (on dried substance)
Physical Form Crystalline Powder
Formula Weight 203.67
Quantity 100g
Color White to almost white
Specific Rotation Condition −47° (20°C c=2,H2O)
Melting Point 140° to 145°C
Merck Index 15, 7398
Specific Rotation -43° to -47° (20°C, 589nm) (c=2, H2O) on dried substance
Molecular Formula C9H13NO2·HCl
Chemical Name or Material L(-)-Phenylephrine hydrochloride
Assay Percent Range 99%
Linear Formula HOC6H4CH(CH2NHCH3)OH·HCl
MDL Number MFCD00012605
CAS 61-76-7
Synonym 3-Hydroxy-α-(methylaminomethyl)benzyl alcohol
Loss on Drying 1% max. (1 g, 105°C)
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material L(-)-Phenylephrine hydrochloride
Assay ≥98.5% (on dried substance)
Assay Percent Range 99%
Molecular Formula C9H13NO2·HCl
Linear Formula HOC6H4CH(CH2NHCH3)OH·HCl
Formula Weight 203.67
MDL Number MFCD00012605
Synonym 3-Hydroxy-α-(methylaminomethyl)benzyl alcohol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents