missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Phenylalanine benzyl ester p-toluenesulfonate, 95%

$37.32 - $145.88

Chemical Identifiers

CAS 1738-78-9
Molecular Formula C23H25NO5S
Molecular Weight (g/mol) 427.515
MDL Number MFCD00035105
InChI Key ZLZGBBIPWXUQST-RSAXXLAASA-N
Synonym h-phe-obzl p-tosylate, l-phenylalanine benzyl ester p-toluenesulfonate, 3-phenyl-l-alanine benzyl ester 4-toluenesulphonate, h-phe-obzl.tosoh, l-phenylalanine benzyl ester tosylate, s-benzyl 2-amino-3-phenylpropanoate 4-methylbenzenesulfonate, o-benzyl-3-phenyl-l-alanine toluene-4-sulphonate, benzyl 2s-2-amino-3-phenylpropanoate; para-toluene sulfonate, h-phe-obzl tos, h-phe-obzl. tos
PubChem CID 12636207
IUPAC Name benzyl (2S)-2-amino-3-phenylpropanoate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)CC(C(=O)OCC2=CC=CC=C2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6270006 View Documents Alfa Aesar™
H6270006
5g Each for $37.32
 
AAH6270014 View Documents Alfa Aesar™
H6270014
25g Each for $145.88
 
Specifications

Chemical Identifiers

1738-78-9
427.515
ZLZGBBIPWXUQST-RSAXXLAASA-N
12636207
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)CC(C(=O)OCC2=CC=CC=C2)N
C23H25NO5S
MFCD00035105
h-phe-obzl p-tosylate, l-phenylalanine benzyl ester p-toluenesulfonate, 3-phenyl-l-alanine benzyl ester 4-toluenesulphonate, h-phe-obzl.tosoh, l-phenylalanine benzyl ester tosylate, s-benzyl 2-amino-3-phenylpropanoate 4-methylbenzenesulfonate, o-benzyl-3-phenyl-l-alanine toluene-4-sulphonate, benzyl 2s-2-amino-3-phenylpropanoate; para-toluene sulfonate, h-phe-obzl tos, h-phe-obzl. tos
benzyl (2S)-2-amino-3-phenylpropanoate;4-methylbenzenesulfonic acid

Specifications

1738-78-9
MFCD00035105
ZLZGBBIPWXUQST-RSAXXLAASA-N
benzyl (2S)-2-amino-3-phenylpropanoate;4-methylbenzenesulfonic acid
12636207
95%
L-Phenylalanine benzyl ester p-toluenesulfonate
C23H25NO5S
h-phe-obzl p-tosylate, l-phenylalanine benzyl ester p-toluenesulfonate, 3-phenyl-l-alanine benzyl ester 4-toluenesulphonate, h-phe-obzl.tosoh, l-phenylalanine benzyl ester tosylate, s-benzyl 2-amino-3-phenylpropanoate 4-methylbenzenesulfonate, o-benzyl-3-phenyl-l-alanine toluene-4-sulphonate, benzyl 2s-2-amino-3-phenylpropanoate; para-toluene sulfonate, h-phe-obzl tos, h-phe-obzl. tos
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)CC(C(=O)OCC2=CC=CC=C2)N
427.515
427.51
5g
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning