Please login to your online account to display your discounted pricing

Alfa Aesar™ L-(+)-Penicillamine, 98+%

$27.43 - $68.86

Chemical Identifiers

Chemical Name or Material L-(+)-Penicillamine
CAS 1113-41-3
Molecular Formula C5H11NO2S
Formula Weight 149.21
MDL Number MFCD00064303
Beilstein 1722374
Merck Index 14,7088
Synonym 3,3-Dimethyl-L-cysteine; 3-Mercapto-L-valine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1392303

  SDS
alfa aesar™
L1392303
1g Each for $68.86

AAL13923MD

  SDS
alfa aesar™
L13923MD
250mg Each for $27.43
Description & Specifications

Specifications

Formula Weight 149.21
MDL Number MFCD00064303
Molecular Formula C5H11NO2S
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Synonym 3,3-Dimethyl-L-cysteine; 3-Mercapto-L-valine
Chemical Name or Material L-(+)-Penicillamine
CAS 1113-41-3
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Merck Index 14,7088
Assay ≥98%
RTECS Number YV9445500
Health Hazard 1 Warning
Recommended Storage 2° to 8°C
Quantity 1g
Melting Point ≈200°C
Optical Rotation +63° (c=1 in 1M NaOH)
Odor Unpleasant
Beilstein 1722374
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C