missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-(+)-Penicillamine, 98+%

Manufacturer:  Alfa Aesar™ L13923MD

 View more versions of this product

Catalog No. AAL13923MD



Specifications

Specifications

L-(+)-Penicillamine
+63° (c=1 in 1M NaOH)
250mg
C5H11NO2S
1722374
l-penicillamine, 3-mercapto-l-valine, penicillamine l-form, l-+-beta-mercaptovaline, l-pen, 2r-2-amino-3-methyl-3-sulfanylbutanoic acid, 3-sulfanyl-l-valine, h-pen-oh, unii-no1yib7oig, 3,3-dimethyl-l-cysteine
CC(C)(C(C(=O)O)N)S
149.208
CHEBI:50869
≥98%
Unpleasant
~200°C
1113-41-3
MFCD00064303
14,7088
VVNCNSJFMMFHPL-GSVOUGTGSA-N
(2R)-2-amino-3-methyl-3-sulfanylbutanoic acid
92863
149.21

Chemical Identifiers

1113-41-3
149.208
VVNCNSJFMMFHPL-GSVOUGTGSA-N
92863
(2R)-2-amino-3-methyl-3-sulfanylbutanoic acid
C5H11NO2S
MFCD00064303
l-penicillamine, 3-mercapto-l-valine, penicillamine l-form, l-+-beta-mercaptovaline, l-pen, 2r-2-amino-3-methyl-3-sulfanylbutanoic acid, 3-sulfanyl-l-valine, h-pen-oh, unii-no1yib7oig, 3,3-dimethyl-l-cysteine
CHEBI:50869
CC(C)(C(C(=O)O)N)S
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : YV9445500

Recommended Storage : 2° to 8°C

Documents