missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-(+)-Pantolactone, 98%

Manufacturer:  Alfa Aesar™ H5582203

 View more versions of this product

Catalog No. AAH5582203



Description

Description

Applications
L-(+)-Pantolactone, is used as Pharmaceutical material and intermediate. It is used as an Intermediate in organic syntheses.

Solubility
Soluble in water, hardly soluble in alcohol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Heat sensitive.
Specifications

Specifications

L-(+)-Pantolactone
120°C to 122°C (15mmHg)
1g
C6H10O3
4659425
SERHXTVXHNVDKA-SCSAIBSYSA-N
(3S)-3-hydroxy-4,4-dimethyloxolan-2-one
736053
98%
+51° (c=3 in Water)
89°C to 94°C
5405-40-3
MFCD00216625
2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3s
CC1(COC(=O)C1O)C
130.143
130.14

Chemical Identifiers

5405-40-3
130.143
SERHXTVXHNVDKA-SCSAIBSYSA-N
736053
CC1(COC(=O)C1O)C
C6H10O3
MFCD00216625
2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3s
(3S)-3-hydroxy-4,4-dimethyloxolan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

Recommended Storage : 2° to 8°C

Documents