missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Ornithine methyl ester dihydrochloride, 97%, Thermo Scientific Chemicals
$570.99 - $570.99
Chemical Identifiers
CAS | 40216-82-8 |
---|---|
Molecular Formula | C6H16Cl2N2O2 |
Molecular Weight (g/mol) | 219.106 |
MDL Number | MFCD00190977 |
InChI Key | SNFZNVWAQDVFAK-XRIGFGBMSA-N |
PubChem CID | 14273060 |
IUPAC Name | methyl (2S)-2,5-diaminopentanoate;dihydrochloride |
SMILES | COC(=O)C(CCCN)N.Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6322522
|
Thermo Scientific Chemicals
H6322522 |
100 g |
Each for $570.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40216-82-8 | |
219.106 | |
SNFZNVWAQDVFAK-XRIGFGBMSA-N | |
methyl (2S)-2,5-diaminopentanoate;dihydrochloride |
C6H16Cl2N2O2 | |
MFCD00190977 | |
14273060 | |
COC(=O)C(CCCN)N.Cl.Cl |
Specifications
40216-82-8 | |
C6H16Cl2N2O2 | |
h-orn-ome.2hcl, l-ornithine methyl ester dihydrochloride, methyl l-ornithine dihydrochloride, s-methyl 2,5-diaminopentanoate dihydrochloride, h-orn-ome?hcl, methyl l-ornithine hcl, h-orn-ome 2hcl, h-orn-ome2hcl, l-ornithine, methyl ester, dihydrochloride, h-orn-ome 2 hcl | |
COC(=O)C(CCCN)N.Cl.Cl | |
219.106 | |
219.11 | |
100 g | |
L-Ornithine methyl ester dihydrochloride |
White | |
MFCD00190977 | |
SNFZNVWAQDVFAK-XRIGFGBMSA-N | |
methyl (2S)-2,5-diaminopentanoate;dihydrochloride | |
14273060 | |
97% | |
Powder |
Safety and Handling
EINECSNumber : 254-841-5
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only