missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Norvalyl diphenylborinate, 95%

Manufacturer:  Alfa Aesar™ H5536803

 View more versions of this product

Catalog No. AAH5536803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

L-Norvalyl diphenylborinate
17093-22-0
MFCD22125141
XAVUFSLMIUPEFM-INIZCTEOSA-N
diphenylboranyl (2S)-2-aminopentanoate
99738133
95%
1g
C17H20BNO2
diphenylboranyl 2s-2-aminopentanoate
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CCC)N
281.162
281.16

Chemical Identifiers

17093-22-0
281.162
XAVUFSLMIUPEFM-INIZCTEOSA-N
99738133
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CCC)N
C17H20BNO2
MFCD22125141
diphenylboranyl 2s-2-aminopentanoate
diphenylboranyl (2S)-2-aminopentanoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents