missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Norvaline methyl ester hydrochloride, 97%

$29.45 - $454.92

Chemical Identifiers

CAS 56558-30-6
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.633
MDL Number MFCD00190974
InChI Key IODNYRVZHKRFIW-JEDNCBNOSA-N
Synonym s-methyl 2-aminopentanoate hydrochloride, l-norvaline methyl ester hydrochloride, methyl l-norvalinate hcl, h-nva-ome.hcl, h-nva-ome hcl, methyl l-norvalinate hydrochloride, s-2-amino-pentanoic acid methyl ester hydrochloride, s-methyl 2-aminopentanoate, methyl 2s-2-aminopentanoate hydrochloride, h-nva-ome?cl
PubChem CID 14118488
IUPAC Name methyl (2S)-2-aminopentanoate;hydrochloride
SMILES CCCC(C(=O)OC)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6501703 View Documents Alfa Aesar™
H6501703
1g Each for $29.45
 
AAH6501706 View Documents Alfa Aesar™
H6501706
5g Each for $114.12
 
AAH6501714 View Documents Alfa Aesar™
H6501714
25g Each for $454.92
 
Specifications

Chemical Identifiers

56558-30-6
167.633
IODNYRVZHKRFIW-JEDNCBNOSA-N
14118488
CCCC(C(=O)OC)N.Cl
C6H14ClNO2
MFCD00190974
s-methyl 2-aminopentanoate hydrochloride, l-norvaline methyl ester hydrochloride, methyl l-norvalinate hcl, h-nva-ome.hcl, h-nva-ome hcl, methyl l-norvalinate hydrochloride, s-2-amino-pentanoic acid methyl ester hydrochloride, s-methyl 2-aminopentanoate, methyl 2s-2-aminopentanoate hydrochloride, h-nva-ome?cl
methyl (2S)-2-aminopentanoate;hydrochloride

Specifications

56558-30-6
MFCD00190974
s-methyl 2-aminopentanoate hydrochloride, l-norvaline methyl ester hydrochloride, methyl l-norvalinate hcl, h-nva-ome.hcl, h-nva-ome hcl, methyl l-norvalinate hydrochloride, s-2-amino-pentanoic acid methyl ester hydrochloride, s-methyl 2-aminopentanoate, methyl 2s-2-aminopentanoate hydrochloride, h-nva-ome?cl
CCCC(C(=O)OC)N.Cl
167.633
167.63
97%
1g
C6H14ClNO2
3561027
IODNYRVZHKRFIW-JEDNCBNOSA-N
methyl (2S)-2-aminopentanoate;hydrochloride
14118488
Solid
White
L-Norvaline methyl ester hydrochloride