missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Norvaline methyl ester hydrochloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6501714
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
L-Norvaline methyl ester hydrochloride | |
White | |
MFCD00190974 | |
s-methyl 2-aminopentanoate hydrochloride, l-norvaline methyl ester hydrochloride, methyl l-norvalinate hcl, h-nva-ome.hcl, h-nva-ome hcl, methyl l-norvalinate hydrochloride, s-2-amino-pentanoic acid methyl ester hydrochloride, s-methyl 2-aminopentanoate, methyl 2s-2-aminopentanoate hydrochloride, h-nva-ome?cl | |
Cl.CCC[C@H](N)C(=O)OC | |
167.63 | |
167.63 | |
25 g |
56558-30-6 | |
C6H14ClNO2 | |
3561027 | |
IODNYRVZHKRFIW-JEDNCBNOSA-N | |
methyl (2S)-2-aminopentanoate hydrochloride | |
14118488 | |
97% | |
Solid |
Chemical Identifiers
56558-30-6 | |
167.63 | |
IODNYRVZHKRFIW-JEDNCBNOSA-N | |
14118488 | |
Cl.CCC[C@H](N)C(=O)OC |
C6H14ClNO2 | |
MFCD00190974 | |
s-methyl 2-aminopentanoate hydrochloride, l-norvaline methyl ester hydrochloride, methyl l-norvalinate hcl, h-nva-ome.hcl, h-nva-ome hcl, methyl l-norvalinate hydrochloride, s-2-amino-pentanoic acid methyl ester hydrochloride, s-methyl 2-aminopentanoate, methyl 2s-2-aminopentanoate hydrochloride, h-nva-ome?cl | |
methyl (2S)-2-aminopentanoate hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C