missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
L-Norleucine | |
White | |
MFCD00064423 | |
l-norleucine, norleucine, h-nle-oh, s-2-aminohexanoic acid, caprine, glycoleucine, l-+-norleucine, l-2-aminohexanoic acid, 2-aminocaproic acid, l-2-aminohexanoate | |
CCCC[C@H](N)C(O)=O | |
131.18 | |
CHEBI:18347 | |
≥99.0% (T) |
327-57-1 | |
C6H13NO2 | |
5 g | |
LRQKBLKVPFOOQJ-YFKPBYRVSA-N | |
(2S)-2-aminohexanoic acid | |
21236 | |
131.18 | |
Crystalline Powder |
Chemical Identifiers
327-57-1 | |
131.18 | |
LRQKBLKVPFOOQJ-YFKPBYRVSA-N | |
21236 | |
(2S)-2-aminohexanoic acid |
C6H13NO2 | |
MFCD00064423 | |
l-norleucine, norleucine, h-nle-oh, s-2-aminohexanoic acid, caprine, glycoleucine, l-+-norleucine, l-2-aminohexanoic acid, 2-aminocaproic acid, l-2-aminohexanoate | |
CHEBI:18347 | |
CCCC[C@H](N)C(O)=O |
Safety and Handling
RTECSNumber : RC6308000
TSCA : Yes