missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Norleucine methyl ester hydrochloride, 98%

$36.09 - $592.18

Chemical Identifiers

CAS 3844-54-0
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00077151
InChI Key FMMOVZRXLNVNBI-RGMNGODLSA-N
Synonym s-methyl 2-aminohexanoate hydrochloride, h-nle-ome hcl, l-norleucine methyl ester hydrochloride, norleucine, methyl ester, hydrochloride, methyl 2s-2-aminohexanoate hydrochloride, h-nle-ome.hcl, h-nle-ome?cl, h-nle-ome hydrochloride, l-norleucine methyl ester hcl, methyl l-norleucinate hydrochloride
PubChem CID 14039065
IUPAC Name methyl (2S)-2-aminohexanoate;hydrochloride
SMILES CCCCC(C(=O)OC)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6587303 View Documents Alfa Aesar™
H6587303
1g Each for $36.09
 
AAH6587306 View Documents Alfa Aesar™
H6587306
5g Each for $142.96
 
AAH6587314 View Documents Alfa Aesar™
H6587314
25g Each for $592.18
 
Specifications

Chemical Identifiers

3844-54-0
181.66
FMMOVZRXLNVNBI-RGMNGODLSA-N
14039065
CCCCC(C(=O)OC)N.Cl
C7H16ClNO2
MFCD00077151
s-methyl 2-aminohexanoate hydrochloride, h-nle-ome hcl, l-norleucine methyl ester hydrochloride, norleucine, methyl ester, hydrochloride, methyl 2s-2-aminohexanoate hydrochloride, h-nle-ome.hcl, h-nle-ome?cl, h-nle-ome hydrochloride, l-norleucine methyl ester hcl, methyl l-norleucinate hydrochloride
methyl (2S)-2-aminohexanoate;hydrochloride

Specifications

3844-54-0
MFCD00077151
s-methyl 2-aminohexanoate hydrochloride, h-nle-ome hcl, l-norleucine methyl ester hydrochloride, norleucine, methyl ester, hydrochloride, methyl 2s-2-aminohexanoate hydrochloride, h-nle-ome.hcl, h-nle-ome?cl, h-nle-ome hydrochloride, l-norleucine methyl ester hcl, methyl l-norleucinate hydrochloride
CCCCC(C(=O)OC)N.Cl
181.66
181.66
98%
L-Norleucine methyl ester hydrochloride
C7H16ClNO2
4880001
FMMOVZRXLNVNBI-RGMNGODLSA-N
methyl (2S)-2-aminohexanoate;hydrochloride
14039065
Solid
1g