missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L(+)-Norleucine, ≥99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC226040500
Specifications
L(+)-Norleucine | |
99+% | |
Authentic | |
04, 432 | |
15, 6795 | |
2-aminocaproic acid, caprine, glycoleucine, h-nle-oh, l-+-norleucine, l-2-aminohexanoate, l-2-aminohexanoic acid, l-norleucine, norleucine, s-2-aminohexanoic acid | |
LRQKBLKVPFOOQJ-YFKPBYRVSA-N | |
(2S)-2-aminohexanoic acid | |
21236 | |
131.17 | |
Crystalline Powder | |
≥99% | |
0275.0°C | |
White | |
50g |
327-57-1 | |
C6H13NO2 | |
MFCD00064423 | |
1% max. (105°C, 4 hrs) | |
(TLC) not detectable | |
Glass bottle | |
CCCCC(C(=O)O)N | |
131.175 | |
CHEBI:18347 | |
+ 22.00 | |
Solubility in water: 1.6 g/100mL (23 c). | |
0.5% max. | |
+22° (20°C c=2,6N HCl) | |
>300.0°C |
Chemical Identifiers
327-57-1 | |
131.175 | |
LRQKBLKVPFOOQJ-YFKPBYRVSA-N | |
21236 | |
(2S)-2-aminohexanoic acid |
C6H13NO2 | |
MFCD00064423 | |
2-aminocaproic acid, caprine, glycoleucine, h-nle-oh, l-+-norleucine, l-2-aminohexanoate, l-2-aminohexanoic acid, l-norleucine, norleucine, s-2-aminohexanoic acid | |
CHEBI:18347 | |
CCCCC(C(=O)O)N |