missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Noradrenaline, 98%

$385.95 - $385.95

Chemical Identifiers

CAS 51-41-2
Molecular Formula C8H11NO3
Formula Weight 169.18
MDL Number MFCD00025592
UN Number UN2811
Beilstein 4231961
Merck Index 14,6695
Synonym L-Arterenol; L-Norepinephrine
Chemical Name or Material L-Noradrenaline
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0808703 SDS Alfa Aesar™
L0808703
1g Each for $385.95
Specifications

Description

Applications
L-Noradrenaline serves as an adrenergic neurotransmitter. It acts as demethylated precursor of epinephrine (adrenaline). It is also used to treat people having low blood pressure, as a sympathomimetic drug to treat people in vasodilatory shock states such as septic shock and neurogenic shock. Further, it is used in the preparation of1,2,3,4-tetrahydro-4,6,7-isoquinolinetriol hydrochloride. In addition to this, it is used in the treatment of hypotension, depression, schizophrenia and attention hyperactivity disorders.

Solubility
Soluble in alkali and dilute hydrochloric acid. Slightly soluble in water, ethanol and diethyl ether.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Light sensitive. Incompatible with acids, bases and oxidizing agents.
Description & Specifications

Chemical Identifiers

51-41-2
169.18
UN2811
14,6695
L-Noradrenaline
C8H11NO3
MFCD00025592
4231961
L-Arterenol; L-Norepinephrine

Specifications

−45° (c=5 in 0.5N HCl)
1g
C8H11NO3
MFCD00025592
4231961
L-Arterenol; L-Norepinephrine
98%
≈220°C
51-41-2
169.18
UN2811
14,6695
L-Noradrenaline

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : DN5950000

Recommended Storage : 2° to 8°C