Please login to your online account to display your discounted pricing

L-NMMA acetate, Tocris

Non-selective NOS inhibitor

$88.05 - $323.08

Chemical Identifiers

Chemical Name or Material L-NMMA acetate
Assay Percent Range >98%
Molecular Formula C7H16N4O2·CH3CO2H
Formula Weight 248.28
Synonym NG-Monomethyl-L-arginine acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

077110

 
r & d systems
RDS077110
10mg Each for $88.05

077150

 
r & d systems
RDS077150
50mg Each for $323.08
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 248.28
Synonym NG-Monomethyl-L-arginine acetate
Chemical Name or Material L-NMMA acetate
CAS 53308-83-1
Molecular Formula C7H16N4O2·CH3CO2H
Assay Percent Range >98%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature