Please login to your online account to display your discounted pricing

L-NIO dihydrochloride, Tocris


Potent eNOS inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS054610

 View more versions of this product

Catalog No. 054610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 246.13
Chemical Name or Material L-NIO dihydrochloride
Assay Percent Range >98%
Molecular Formula C7H15N3O2·2HCl
CAS 159190-44-0
Synonym N5-(1-Iminoethyl)-L-ornithine dihydrochloride
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material L-NIO dihydrochloride
Assay Percent Range >98%
Molecular Formula C7H15N3O2·2HCl
Formula Weight 246.13
Synonym N5-(1-Iminoethyl)-L-ornithine dihydrochloride
Accessories

Desiccate at -20°C