Please login to your online account to display your discounted pricing

L-NIL hydrochloride, Tocris

Selective iNOS inhibitor

$164.61 - $651.02

Chemical Identifiers

Chemical Name or Material L-NIL hydrochloride
Assay Percent Range >98%
Molecular Formula C8H17N3O2·HCl
Formula Weight 223.7
Synonym N6-(1-Iminoethyl)-L-lysine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

113950

 
r & d systems
RDS113950
50mg Each for $651.02

113910

 
r & d systems
RDS113910
10mg Each for $164.61
Description & Specifications

Specifications

Chemical Name or Material L-NIL hydrochloride
CAS 150403-89-7
Recommended Storage Desiccate at -20°C
Assay Percent Range >98%
Molecular Formula C8H17N3O2·HCl
Formula Weight 223.7
Synonym N6-(1-Iminoethyl)-L-lysine hydrochloride
Quantity 50mg
Safety and Handling

Desiccate at -20°C