Please login to your online account to display your discounted pricing

Alfa Aesar™ L-myo-Inositol-1,4,5-trisphosphate sodium salt, 98%


Quantity 0.1mg

Manufacturer: alfa aesar™  J67018EXZ

 View more versions of this product

Catalog No. AAJ67018EXZ

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 486.04
CAS 2068-89-5
Chemical Name or Material L-myo-Inositol-1,4,5-trisphosphate sodium salt
Assay 98%
Synonym 1,4,5-IP3 sodium salt; Ins(1,4,5)P3 sodium salt
Storage Note 1 -30° to -10°C
Color White to off-white
Quantity 0.1mg
Physical Form Lyophilized Powder
MDL Number MFCD18427941

Specifications

Chemical Name or Material L-myo-Inositol-1,4,5-trisphosphate sodium salt
CAS 2068-89-5
Formula Weight 486.04
MDL Number MFCD18427941
Synonym 1,4,5-IP3 sodium salt; Ins(1,4,5)P3 sodium salt
Accessories

-30° to -10°C