Please login to your online account to display your discounted pricing

Alfa Aesar™ L-myo-Inositol-1,4,5-trisphosphate sodium salt, 98%

Chemical Identifiers

Chemical Name or Material L-myo-Inositol-1,4,5-trisphosphate sodium salt
CAS 2068-89-5
Formula Weight 486.04
MDL Number MFCD18427941
Synonym 1,4,5-IP3 sodium salt; Ins(1,4,5)P3 sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67018EXZ

 
alfa aesar™
J67018EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 486.04
Chemical Name or Material L-myo-Inositol-1,4,5-trisphosphate sodium salt
CAS 2068-89-5
Assay 98%
Synonym 1,4,5-IP3 sodium salt; Ins(1,4,5)P3 sodium salt
Physical Form Lyophilized Powder
MDL Number MFCD18427941
Color White to off-white
Quantity 0.1mg
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C