Please login to your online account to display your discounted pricing

Alfa Aesar™ L-(-)-Methioninol, 97%

$43.73 - $118.84

Chemical Identifiers

Chemical Name or Material L-(-)-Methioninol
CAS 2899-37-8
Molecular Formula C5H13NOS
Formula Weight 135.23
MDL Number MFCD00004735
Beilstein 2231656
Synonym (S)-(-)-2-Amino-4-methylthio-1-butanol; H-Met-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2421103

 
alfa aesar™
B2421103
1g Each for $118.84

AAB24211MD

 
alfa aesar™
B24211MD
250mg Each for $43.73
Description & Specifications

Specifications

Formula Weight 135.23
Flash Point >110°C (230°F)
Optical Rotation −21° (c=5 in Water)
MDL Number MFCD00004735
Melting Point 31° to 33°C
Chemical Name or Material L-(-)-Methioninol
Quantity 1g
Assay 97%
Molecular Formula C5H13NOS
CAS 2899-37-8
Beilstein 2231656
Synonym (S)-(-)-2-Amino-4-methylthio-1-butanol; H-Met-ol