missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Methionine tert-butyl ester hydrochloride, 95%

$54.74 - $649.23

Chemical Identifiers

CAS 91183-71-0
Molecular Formula C9H20ClNO2S
Molecular Weight (g/mol) 241.774
MDL Number MFCD00038896
InChI Key KZJQROCWHZGZBJ-FJXQXJEOSA-N
Synonym h-met-otbu.hcl, h-met-otbu hcl, h-met-otbu hydrochloride, s-tert-butyl 2-amino-4-methylthio butanoate hydrochloride, l-methionine tert-butyl ester hydrochloride, tert-butyl l-methioninate-hydrogen chloride 1/1, l-methionine, 1,1-dimethylethyl ester, hydrochloride, 2-amino-4-methylsulfanyl-butyric acid tert-bu ester, hydrochloride, tert-butyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-otbu?hci
PubChem CID 13141795
IUPAC Name tert-butyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
SMILES CC(C)(C)OC(=O)C(CCSC)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65439MD View Documents Alfa Aesar™
H65439MD
250mg Each for $54.74
 
AAH6543903 View Documents Alfa Aesar™
H6543903
1g Each for $163.46
 
AAH6543906 View Documents Alfa Aesar™
H6543906
5g Each for $649.23
 
Description

Description

Applications
L-Methionine tert-butyl ester hydrochloride′s main function is to act as the primary “Start” sequence on mRNA so that the ribosomes can start translating the mRNA into proteins.

Notes
Store at 4°C. Incompatible with heat and oxidizing agents.
Specifications

Chemical Identifiers

91183-71-0
241.774
KZJQROCWHZGZBJ-FJXQXJEOSA-N
13141795
CC(C)(C)OC(=O)C(CCSC)N.Cl
C9H20ClNO2S
MFCD00038896
h-met-otbu.hcl, h-met-otbu hcl, h-met-otbu hydrochloride, s-tert-butyl 2-amino-4-methylthio butanoate hydrochloride, l-methionine tert-butyl ester hydrochloride, tert-butyl l-methioninate-hydrogen chloride 1/1, l-methionine, 1,1-dimethylethyl ester, hydrochloride, 2-amino-4-methylsulfanyl-butyric acid tert-bu ester, hydrochloride, tert-butyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-otbu?hci
tert-butyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride

Specifications

91183-71-0
MFCD00038896
h-met-otbu.hcl, h-met-otbu hcl, h-met-otbu hydrochloride, s-tert-butyl 2-amino-4-methylthio butanoate hydrochloride, l-methionine tert-butyl ester hydrochloride, tert-butyl l-methioninate-hydrogen chloride 1/1, l-methionine, 1,1-dimethylethyl ester, hydrochloride, 2-amino-4-methylsulfanyl-butyric acid tert-bu ester, hydrochloride, tert-butyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-otbu?hci
CC(C)(C)OC(=O)C(CCSC)N.Cl
241.774
241.77
250mg
C9H20ClNO2S
4884935
KZJQROCWHZGZBJ-FJXQXJEOSA-N
tert-butyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
13141795
95%
L-Methionine tert-butyl ester hydrochloride
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C