missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Methionine tert-butyl ester hydrochloride, 95%

Manufacturer:  Alfa Aesar™ H6543906

 View more versions of this product

Catalog No. AAH6543906



Description

Description

Applications
L-Methionine tert-butyl ester hydrochloride′s main function is to act as the primary “Start” sequence on mRNA so that the ribosomes can start translating the mRNA into proteins.

Notes
Store at 4°C. Incompatible with heat and oxidizing agents.
Specifications

Specifications

L-Methionine tert-butyl ester hydrochloride
C9H20ClNO2S
4884935
KZJQROCWHZGZBJ-FJXQXJEOSA-N
tert-butyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
13141795
95%
91183-71-0
MFCD00038896
h-met-otbu.hcl, h-met-otbu hcl, h-met-otbu hydrochloride, s-tert-butyl 2-amino-4-methylthio butanoate hydrochloride, l-methionine tert-butyl ester hydrochloride, tert-butyl l-methioninate-hydrogen chloride 1/1, l-methionine, 1,1-dimethylethyl ester, hydrochloride, 2-amino-4-methylsulfanyl-butyric acid tert-bu ester, hydrochloride, tert-butyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-otbu?hci
CC(C)(C)OC(=O)C(CCSC)N.Cl
241.774
241.77
5g

Chemical Identifiers

91183-71-0
241.774
KZJQROCWHZGZBJ-FJXQXJEOSA-N
13141795
CC(C)(C)OC(=O)C(CCSC)N.Cl
C9H20ClNO2S
MFCD00038896
h-met-otbu.hcl, h-met-otbu hcl, h-met-otbu hydrochloride, s-tert-butyl 2-amino-4-methylthio butanoate hydrochloride, l-methionine tert-butyl ester hydrochloride, tert-butyl l-methioninate-hydrogen chloride 1/1, l-methionine, 1,1-dimethylethyl ester, hydrochloride, 2-amino-4-methylsulfanyl-butyric acid tert-bu ester, hydrochloride, tert-butyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-otbu?hci
tert-butyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

SDS Documents