missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals L-Methionine sulfone, 98+%
Supplier: Thermo Scientific Chemicals A1702706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
254°C (decomposition) | |
7314-32-1 | |
MFCD00066020 | |
l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+% | |
CS(=O)(=O)CC[C@H](N)C(O)=O | |
181.21 | |
CHEBI:21363 | |
≥99% | |
+11.6° (c=0.69 in Water) |
L-Methionine sulfone | |
C5H11NO4S | |
1725510 | |
UCUNFLYVYCGDHP-BYPYZUCNSA-N | |
(2S)-2-amino-4-methylsulfonylbutanoic acid | |
445282 | |
181.21 | |
5 g |
Chemical Identifiers
7314-32-1 | |
181.21 | |
UCUNFLYVYCGDHP-BYPYZUCNSA-N | |
445282 | |
(2S)-2-amino-4-methylsulfonylbutanoic acid |
C5H11NO4S | |
MFCD00066020 | |
l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+% | |
CHEBI:21363 | |
CS(=O)(=O)CC[C@H](N)C(O)=O |
Safety and Handling
EINECSNumber : 230-774-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only