missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Methionine sulfone, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC439000010

 View more versions of this product

Catalog No. AC439000010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

L-Methionine sulfone
Authentic
12.7
White
1g
98%
2s-2-amino-4-methanesulfonylbutanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, h-met o 2-oh, l-2-amino-4-methylsulfonyl butanoic acid, l-methionine sulfone, l-methioninesulfone,99+%, methionine s,s-dioxide, s-2-amino-4-methylsulfonyl butanoic acid, s-amino-4-methylsulphonyl butyric acid, s-dioxymethionine
CS(=O)(=O)CCC(C(=O)O)N
181.21
CHEBI:21363
Powder
32.31 to 33.97% C, 7.54 to 7.92% N
Glass Bottle
1% Max. (K.F.)
242.3°C to 243.1°C
7314-32-1
C5H11NO4S
UCUNFLYVYCGDHP-BYPYZUCNSA-N
(2S)-2-amino-4-methylsulfonylbutanoic acid
445282
181.21
98%

Chemical Identifiers

7314-32-1
181.21
2s-2-amino-4-methanesulfonylbutanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, h-met o 2-oh, l-2-amino-4-methylsulfonyl butanoic acid, l-methionine sulfone, l-methioninesulfone,99+%, methionine s,s-dioxide, s-2-amino-4-methylsulfonyl butanoic acid, s-amino-4-methylsulphonyl butyric acid, s-dioxymethionine
CHEBI:21363
CS(=O)(=O)CCC(C(=O)O)N
C5H11NO4S
UCUNFLYVYCGDHP-BYPYZUCNSA-N
445282
(2S)-2-amino-4-methylsulfonylbutanoic acid
Safety and Handling

Safety and Handling

EINECSNumber : 230-774-7

SDS Documents