missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Methionine sulfone, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 7314-32-1 |
---|---|
Molecular Formula | C5H11NO4S |
Molecular Weight (g/mol) | 181.21 |
InChI Key | UCUNFLYVYCGDHP-BYPYZUCNSA-N |
Synonym | l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+% |
PubChem CID | 445282 |
ChEBI | CHEBI:21363 |
IUPAC Name | (2S)-2-amino-4-methylsulfonylbutanoic acid |
SMILES | CS(=O)(=O)CCC(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC439000050
|
Thermo Scientific Chemicals
439000050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7314-32-1 | |
181.21 | |
l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+% | |
CHEBI:21363 | |
CS(=O)(=O)CCC(C(=O)O)N |
C5H11NO4S | |
UCUNFLYVYCGDHP-BYPYZUCNSA-N | |
445282 | |
(2S)-2-amino-4-methylsulfonylbutanoic acid |
Specifications
7314-32-1 | |
Authentic | |
C5H11NO4S | |
12.7 | |
UCUNFLYVYCGDHP-BYPYZUCNSA-N | |
(2S)-2-amino-4-methylsulfonylbutanoic acid | |
445282 | |
181.21 | |
L-Methionine sulfone |
32.31 to 33.97% C, 7.54 to 7.92% N | |
98% | |
5g | |
l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+% | |
CS(=O)(=O)CCC(C(=O)O)N | |
181.21 | |
CHEBI:21363 | |
98% |
Safety and Handling
EINECSNumber : 230-774-7
RUO â Research Use Only