missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Methionine sulfone, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 439000050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
L-Methionine sulfone | |
32.31 to 33.97% C, 7.54 to 7.92% N | |
98% | |
5g | |
l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+% | |
CS(=O)(=O)CCC(C(=O)O)N | |
181.21 | |
CHEBI:21363 | |
98% |
7314-32-1 | |
Authentic | |
C5H11NO4S | |
12.7 | |
UCUNFLYVYCGDHP-BYPYZUCNSA-N | |
(2S)-2-amino-4-methylsulfonylbutanoic acid | |
445282 | |
181.21 |
Chemical Identifiers
7314-32-1 | |
181.21 | |
l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+% | |
CHEBI:21363 | |
CS(=O)(=O)CCC(C(=O)O)N |
C5H11NO4S | |
UCUNFLYVYCGDHP-BYPYZUCNSA-N | |
445282 | |
(2S)-2-amino-4-methylsulfonylbutanoic acid |
Safety and Handling
EINECSNumber : 230-774-7
RUO â Research Use Only