Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Methionine methylsulfonium iodide, 99%

$45.81 - $516.82

Chemical Identifiers

Chemical Name or Material L-Methionine methylsulfonium iodide
CAS 34236-06-1
Molecular Formula C6H14INO2S
Formula Weight 291.15
MDL Number MFCD00050272
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2134206

  SDS
alfa aesar™
B2134206
5g Each for $45.81

AAB2134222

  SDS
alfa aesar™
B2134222
100g Each for $516.82
Description & Specifications

Specifications

Formula Weight 291.15
Chemical Name or Material L-Methionine methylsulfonium iodide
Assay 99%
Molecular Formula C6H14INO2S
MDL Number MFCD00050272
CAS 34236-06-1
Melting Point 156° to 157°C
Quantity 5g

Applications
L-Methionine methylsulfonium iodide, is used as an pharmaceutical intermediate. It is also widely used in polyelectrolytes in case of polymer studies.

Solubility
Less soluble in water 0.001 mg/ml.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.