missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Methionine Methyl Ester Hydrochloride 96.0+%, TCI America™

Manufacturer:  Tci America M08535G

 View more versions of this product

Catalog No. M08535G



Specifications

Specifications

L-Methionine Methyl Ester Hydrochloride
151°C
2491-18-1
MFCD00012491
MEVUPUNLVKELNV-JEDNCBNOSA-N
methyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
11435579
Crystal-Powder at 20°C
Undesignated
5g
C6H14ClNO2S
h-d-met-ome hydrochloride, h-met-ome hydrochloride, h-met-ome. hcl, h-met-ome.hcl, l-methionine methyl ester hydrochloride, met-ome. hcl, met-ome.hcl, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, pubchem14925
COC(=O)C(CCSC)N.Cl
199.693
199.69
≥96.0% (T)

Chemical Identifiers

2491-18-1
199.693
MEVUPUNLVKELNV-JEDNCBNOSA-N
11435579
COC(=O)C(CCSC)N.Cl
C6H14ClNO2S
MFCD00012491
h-d-met-ome hydrochloride, h-met-ome hydrochloride, h-met-ome. hcl, h-met-ome.hcl, l-methionine methyl ester hydrochloride, met-ome. hcl, met-ome.hcl, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, pubchem14925
methyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents