missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Methionine isopropyl ester hydrochloride, 97%

$43.03 - $422.54

Chemical Identifiers

CAS 85391-05-5
Molecular Formula C8H18ClNO2S
Molecular Weight (g/mol) 227.747
MDL Number MFCD00190954
InChI Key ONXXRAMAPIOLSS-FJXQXJEOSA-N
Synonym h-met-oipr.hcl, h-met-oipr hcl, l-methionine isopropyl ester hydrochloride, isopropyl l-methionate hydrochloride, s-isopropyl 2-amino-4-methylthio butanoate hydrochloride, isopropyl l-methionate hcl, isopropyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-oipr?cl, h-met-oipr ??hcl, methionine isopropyl ester hydrochloride
PubChem CID 21156159
IUPAC Name propan-2-yl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
SMILES CC(C)OC(=O)C(CCSC)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65339MD View Documents Alfa Aesar™
H65339MD
250mg Each for $43.03
 
AAH6533903 View Documents Alfa Aesar™
H6533903
1g Each for $113.35
 
AAH6533906 View Documents Alfa Aesar™
H6533906
5g Each for $422.54
 
Specifications

Chemical Identifiers

85391-05-5
227.747
ONXXRAMAPIOLSS-FJXQXJEOSA-N
21156159
CC(C)OC(=O)C(CCSC)N.Cl
C8H18ClNO2S
MFCD00190954
h-met-oipr.hcl, h-met-oipr hcl, l-methionine isopropyl ester hydrochloride, isopropyl l-methionate hydrochloride, s-isopropyl 2-amino-4-methylthio butanoate hydrochloride, isopropyl l-methionate hcl, isopropyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-oipr?cl, h-met-oipr ??hcl, methionine isopropyl ester hydrochloride
propan-2-yl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride

Specifications

85391-05-5
MFCD00190954
h-met-oipr.hcl, h-met-oipr hcl, l-methionine isopropyl ester hydrochloride, isopropyl l-methionate hydrochloride, s-isopropyl 2-amino-4-methylthio butanoate hydrochloride, isopropyl l-methionate hcl, isopropyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-oipr?cl, h-met-oipr ??hcl, methionine isopropyl ester hydrochloride
CC(C)OC(=O)C(CCSC)N.Cl
227.747
227.75
250mg
C8H18ClNO2S
3912939
ONXXRAMAPIOLSS-FJXQXJEOSA-N
propan-2-yl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
21156159
97%
L-Methionine isopropyl ester hydrochloride
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C