missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Methionine isopropyl ester hydrochloride, 97%

Manufacturer:  Alfa Aesar™ H6533906

 View more versions of this product

Catalog No. AAH6533906



Specifications

Specifications

L-Methionine isopropyl ester hydrochloride
C8H18ClNO2S
3912939
ONXXRAMAPIOLSS-FJXQXJEOSA-N
propan-2-yl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
21156159
97%
85391-05-5
MFCD00190954
h-met-oipr.hcl, h-met-oipr hcl, l-methionine isopropyl ester hydrochloride, isopropyl l-methionate hydrochloride, s-isopropyl 2-amino-4-methylthio butanoate hydrochloride, isopropyl l-methionate hcl, isopropyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-oipr?cl, h-met-oipr ??hcl, methionine isopropyl ester hydrochloride
CC(C)OC(=O)C(CCSC)N.Cl
227.747
227.75
5g

Chemical Identifiers

85391-05-5
227.747
ONXXRAMAPIOLSS-FJXQXJEOSA-N
21156159
CC(C)OC(=O)C(CCSC)N.Cl
C8H18ClNO2S
MFCD00190954
h-met-oipr.hcl, h-met-oipr hcl, l-methionine isopropyl ester hydrochloride, isopropyl l-methionate hydrochloride, s-isopropyl 2-amino-4-methylthio butanoate hydrochloride, isopropyl l-methionate hcl, isopropyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-oipr?cl, h-met-oipr ??hcl, methionine isopropyl ester hydrochloride
propan-2-yl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

SDS Documents