missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Methionine ethyl ester hydrochloride, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC205410250

 View more versions of this product

Catalog No. AC205410250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

L-Methionine ethyl ester hydrochloride
+ 21.00
25g
2899-36-7
C7H16ClNO2S
ethyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride
CCOC(=O)C(CCSC)N.Cl
213.72
213.73
+ 21.00 (25.00°C c=2,EtOH)
82.0°C to 92.0°C
99%
99%
CH3SCH2CH2CH(NH2)CO2C2H5·HCl
KPWCQEUBJAIORR-RGMNGODLSA-N
ethyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
3083785
99%

Chemical Identifiers

2899-36-7
213.72
ethyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride
ethyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
C7H16ClNO2S
KPWCQEUBJAIORR-RGMNGODLSA-N
3083785
CCOC(=O)C(CCSC)N.Cl
Safety and Handling

Safety and Handling

EINECSNumber : 220-787-6

SDS Documents