Please login to your online account to display your discounted pricing

L-Methionine ethyl ester hydrochloride, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material L-Methionine ethyl ester hydrochloride
Name Note 99%
CAS 2899-36-7
Assay Percent Range 99%
Molecular Formula C7H15NO2S·HCl
Linear Formula CH3SCH2CH2CH(NH2)CO2C2H5·HCl
Formula Weight 213.73
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC205410250

  SDS
thermo fisher scientific
AC205410250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC205410100

  SDS
thermo fisher scientific
AC205410100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 213.73
Quantity 25g
EINECS Number 220-787-6
Specific Rotation Condition + 21.00 (25.00°C c=2,EtOH)
Chemical Name or Material L-Methionine ethyl ester hydrochloride
Assay Percent Range 99%
CAS 2899-36-7
Molecular Formula C7H15NO2S·HCl
Name Note 99%
Linear Formula CH3SCH2CH2CH(NH2)CO2C2H5·HCl