missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Methionine ethyl ester hydrochloride, 99%

$49.81 - $396.42

Chemical Identifiers

CAS 2899-36-7
Molecular Formula C7H15NO2S·HCl
Formula Weight 213.73
MDL Number MFCD00012508
Synonym H-Met-OEt·HCl
Chemical Name or Material L-Methionine ethyl ester hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2505506 SDS Alfa Aesar™
B2505506
5g Each for $49.81
AAB2505514 SDS Alfa Aesar™
B2505514
25g Each for $115.04
AAB2505522 SDS Alfa Aesar™
B2505522
100g Each for $396.42
Specifications Description & Specifications

Chemical Identifiers

2899-36-7
213.73
H-Met-OEt·HCl
C7H15NO2S·HCl
MFCD00012508
L-Methionine ethyl ester hydrochloride

Specifications

2899-36-7
213.73
H-Met-OEt·HCl
5g
99%
C7H15NO2S·HCl
MFCD00012508
90° to 92°C
L-Methionine ethyl ester hydrochloride